Tisk…

7-(Dimethylamino)-4-(trifluoromethyl)coumarin 99%

Dodavatel: Apollo Scientific
Varování

PC48809-25G PC48809-5G
APOSPC48809-25GEA 8140 CZK
APOSPC48809-25G APOSPC48809-5G
7-(Dimethylamino)-4-(trifluoromethyl)coumarin 99%
7-(Dimethylamino)-4-(trifluoromethyl)coumarin
Vzorec: C₁₂H₁₀F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD00041867
CAS číslo: 53518-14-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR