Tisk…
{[(1-Cyano-2-ethoxy-2-oxoethylidene)amino]oxy}dimethylamino(morpholin-4-yl)carbenium hexafluorophosphate, Technical Grade
Dodavatel: Apollo Scientific
Varování
Synonyms: [[(E)-(1-cyano-2-ethoxy-2-oxoethylidene)amino]oxy-morpholin-4-ylmethylidene]-dimethylazanium hexafluorophosphate
PC48805-100G PC48805-25G
APOSPC48805-100GEA 0 CZK
APOSPC48805-100G APOSPC48805-25G
{[(1-Cyano-2-ethoxy-2-oxoethylidene)amino]oxy}dimethylamino(morpholin-4-yl)carbenium hexafluorophosphate, Technical Grade
{[(1-Cyano-2-ethoxy-2-oxoethylidene)amino]oxy}dimethylamino(morpholin-4-yl)carbenium hexafluorophosphate
Vzorec: C₁₂H₁₉F₆N₄O₄P
MW: 428.272 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24368564
CAS číslo: 1075198-30-9