1-Bromo-2-(1,1-difluoroethyl)benzene 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC48786-1G
PC48786-25G
PC48786-5G
APOSPC48786-1GEA
2280
CZK
APOSPC48786-1G
APOSPC48786-25G
APOSPC48786-5G
1-Bromo-2-(1,1-difluoroethyl)benzene 97%
1-Bromo-2-(1,1-difluoroethyl)benzene
Vzorec:
C₈H₇BrF₂ Storage Temperature: Ambient |
MDL Number:
MFCD09996896 CAS číslo: 444581-46-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...