Tisk…

Methyl-2,6-difluoro-3-nitrobenzoate

Dodavatel: Apollo Scientific
Varování

Synonyms: Methyl 2,6-difluoro-3-nitrobenzoate

PC48724-25G PC48724-5G PC48724-100G
APOSPC48724-25GEA 2900 CZK
APOSPC48724-25G APOSPC48724-5G APOSPC48724-100G
Methyl-2,6-difluoro-3-nitrobenzoate
Methyl-2,6-difluoro-3-nitrobenzoate
Vzorec: C₈H₅F₂NO₄
MW: 217,128 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368850
CAS číslo: 84832-01-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR