Tisk…
2-Chloro-5-(trifluoromethyl)benzoic acid, Technical Grade
Dodavatel: Apollo Scientific
Synonyms: 2-Chloro-5-trifluoromethylbenzoic acid, 6-Chloro-α,α,α-trifluoro-m-toluic acid
PC48678-25G PC48678-5G
APOSPC48678-25GEA 827 CZK
APOSPC48678-25G APOSPC48678-5G
2-Chloro-5-(trifluoromethyl)benzoic acid, Technical Grade
2-Chloro-5-(trifluoromethyl)benzoic acid
Vzorec: C₈H₄ClF₃O₂
MW: 224.57
Storage Temperature: Ambient
MDL Number: MFCD00061094
CAS číslo: 657-06-7