7-Hydroxy-4-(trifluoromethyl)coumarin 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC4866-1G
PC4866-5G
APOSPC4866-1GEA
662
CZK
APOSPC4866-1G
APOSPC4866-5G
7-Hydroxy-4-(trifluoromethyl)coumarin 97%
7-Hydroxy-4-(trifluoromethyl)coumarin
Vzorec:
C₁₀H₅F₃O₃ Bod tání: 178…188 °C Storage Temperature: Ambient |
MDL Number:
MFCD00037578 CAS číslo: 575-03-1 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...