4-Nitro-3-(trifluoromethyl)phenol 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
α,α,α-Trifluoro-4-nitro-m-cresol, 5-Hydroxy-2-nitrobenzotrifluoride
PC4860-1G
PC4860-25G
PC4860-5G
APOSPC4860-1GEA
622
CZK
APOSPC4860-1G
APOSPC4860-25G
APOSPC4860-5G
4-Nitro-3-(trifluoromethyl)phenol 97%
4-Nitro-3-(trifluoromethyl)phenol
Vzorec:
C₇H₄F₃NO₃ Bod varu: 135…138 °C/0,01mmHg Bod tání: 72…80 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014714 CAS číslo: 88-30-2 EINECS: 201-818-2 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...