Tisk…

4-Bromo-2-fluoro-N-methylbenzamide

Dodavatel: Apollo Scientific

PC48582-1G PC48582-250MG
APOSPC48582-1GEA 8870 CZK
APOSPC48582-1G APOSPC48582-250MG
4-Bromo-2-fluoro-N-methylbenzamide
4-Bromo-2-fluoro-N-methylbenzamide
Vzorec: C₈H₇BrFNO
MW: 232,0497 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09878362
CAS číslo: 749927-69-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR