3-Chloro-2,4,5-trifluorobenzoic acid
Dodavatel: Apollo Scientific
|
Varování
|
PC4854-25G
PC4854-5G
APOSPC4854-25GEA
3110
CZK
APOSPC4854-25G
APOSPC4854-5G
3-Chloro-2,4,5-trifluorobenzoic acid
3-Chloro-2,4,5-trifluorobenzoic acid
Vzorec:
C₇H₂ClF₃O₂ Bod tání: 112…116 °C Storage Temperature: Ambient |
MDL Number:
MFCD00153101 CAS číslo: 101513-77-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...