Tisk…

3-Bromo-4-(trifluoromethyl)benzoic acid 97%

Dodavatel: Apollo Scientific
Varování

PC48524-1G PC48524-25G PC48524-5G
APOSPC48524-1GEA 622 CZK
APOSPC48524-1G APOSPC48524-25G APOSPC48524-5G
3-Bromo-4-(trifluoromethyl)benzoic acid 97%
3-Bromo-4-(trifluoromethyl)benzoic acid
Vzorec: C₈H₄BrF₃O₂
MW: 269,0200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185386
CAS číslo: 581813-17-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR