Tisk…
{[(1-Cyano-2-ethoxy-2-oxoethylidene)amino]oxy}dimethylamino(morpholin-4-yl)carbenium hexafluorophosphate 98%
Dodavatel: Apollo Scientific
Varování
Synonyms: [[(E)-(1-cyano-2-ethoxy-2-oxoethylidene)amino]oxy-morpholin-4-ylmethylidene]-dimethylazanium hexafluorophosphate
PC48468-100G PC48468-25G PC48468-5G
APOSPC48468-100GEA 5790 CZK
APOSPC48468-100G APOSPC48468-25G APOSPC48468-5G
{[(1-Cyano-2-ethoxy-2-oxoethylidene)amino]oxy}dimethylamino(morpholin-4-yl)carbenium hexafluorophosphate 98%
{[(1-Cyano-2-ethoxy-2-oxoethylidene)amino]oxy}dimethylamino(morpholin-4-yl)carbenium hexafluorophosphate
Vzorec: C₁₂H₁₉F₆N₄O₄P
MW: 428,272 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24368564
CAS číslo: 1075198-30-9