Tisk…

2-Amino-N-(2,2,2-trifluoroethyl)acetamide hydrochloride 95%

Dodavatel: Apollo Scientific
Varování

PC48392-1G PC48392-5G
APOSPC48392-1GEA 612 CZK
APOSPC48392-1G APOSPC48392-5G
2-Amino-N-(2,2,2-trifluoroethyl)acetamide hydrochloride 95%
2-Amino-N-(2,2,2-trifluoroethyl)acetamide hydrochloride
Vzorec: C₄H₇N₂OF₃·HCl
Storage Temperature: Refrigerator
MDL Number: MFCD09944516
CAS číslo: 1171331-39-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR