2-Methoxy-3-(trifluoromethyl)benzonitrile 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC48356-1G
PC48356-5G
PC48356-25G
PC48356-100G
APOSPC48356-1GEA
1820
CZK
APOSPC48356-1G
APOSPC48356-5G
APOSPC48356-25G
APOSPC48356-100G
2-Methoxy-3-(trifluoromethyl)benzonitrile 97%
2-Methoxy-3-(trifluoromethyl)benzonitrile
Vzorec:
C₉H₆F₃NO Storage Temperature: Ambient |
MDL Number:
MFCD09832310 CAS číslo: 1017778-62-9 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...