3'-Fluoro-2'-methoxyacetophenone 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1-(3-Fluoro-2-methoxyphenyl)ethanone
PC48354-1G
PC48354-250MG
PC48354-5G
APOSPC48354-1GEA
1990
CZK
APOSPC48354-1G
APOSPC48354-250MG
APOSPC48354-5G
3'-Fluoro-2'-methoxyacetophenone 97%
3'-Fluoro-2'-methoxyacetophenone
Vzorec:
C₉H₉FO₂ MW: 168,17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12545933 CAS číslo: 295779-86-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...