4'-Chloro-5-fluoro-1,1'-biphenyl-2-amine
Dodavatel: Apollo Scientific
Synonyms:
2-Amino-4'-chloro-5-fluorobiphenyl
PC48313-1G
PC48313-100MG
PC48313-250MG
PC48313-500MG
PC48313-2.5G
PC48313-5G
APOSPC48313-1GEA
10420
CZK
APOSPC48313-1G
APOSPC48313-100MG
APOSPC48313-250MG
APOSPC48313-500MG
APOSPC48313-2.5G
APOSPC48313-5G
4'-Chloro-5-fluoro-1,1'-biphenyl-2-amine
4'-Chloro-5-fluoro-1,1'-biphenyl-2-amine
Vzorec:
C₁₂H₉ClFN Storage Temperature: Refrigerator |
MDL Number:
MFCD12824033 CAS číslo: 188731-34-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...