2-Chloro-4-(trifluoromethyl)benzoic acid 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Chloro-4-trifluoromethylbenzoic acid, 2-Chloro-α,α,α-trifluoro-p-toluic acid
PC48310-25G
PC48310-100G
PC48310-5G
APOSPC48310-25GEA
2570
CZK
APOSPC48310-25G
APOSPC48310-100G
APOSPC48310-5G
2-Chloro-4-(trifluoromethyl)benzoic acid 97%
2-Chloro-4-(trifluoromethyl)benzoic acid
Vzorec:
C₈H₄ClF₃O₂ MW: 224.57 Bod tání: 114…117 °C Storage Temperature: Ambient |
MDL Number:
MFCD06208178 CAS číslo: 23228-45-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...