Tisk…

5-Chloro-2-fluoro-4-iodopyridine 97%

Dodavatel: Apollo Scientific

PC48308-25G
APOSPC48308-25GEA 5860 CZK
APOSPC48308-25G
5-Chloro-2-fluoro-4-iodopyridine 97%
5-Chloro-2-fluoro-4-iodopyridine
Vzorec: C₅H₂ClFIN
MW: 257,43 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04112566
CAS číslo: 659731-48-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR