2-Nitro-4-(pentafluorosulphanyl)aniline 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Amino-3-nitrophenylsulphur pentafluoride
PC4815-1G
PC4815-250MG
APOSPC4815-1GEA
27710
CZK
APOSPC4815-1G
APOSPC4815-250MG
2-Nitro-4-(pentafluorosulphanyl)aniline 95%
2-Nitro-4-(pentafluorosulphanyl)aniline
Vzorec:
C₆H₅F₅N₂O₂S MW: 264.17 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11501034 CAS číslo: 197385-06-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...