Tisk…

2-(Difluoromethoxy)phenylacetonitrile 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-(2-(Difluoromethoxy)phenyl)acetonitrile

PC4809-1G PC4809-5G PC4809-10G
APOSPC4809-1GEA 622 CZK
APOSPC4809-1G APOSPC4809-5G APOSPC4809-10G
2-(Difluoromethoxy)phenylacetonitrile 98%
2-(Difluoromethoxy)phenylacetonitrile
Vzorec: C₉H₇F₂NO
Storage Temperature: Ambient
MDL Number: MFCD00236240
CAS číslo: 41429-22-5
UN: 3276
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR