Hexafluoropropene trimer 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC4792-100G
PC4792-25G
APOSPC4792-100GEA
6420
CZK
APOSPC4792-100G
APOSPC4792-25G
Hexafluoropropene trimer 97%
Hexafluoropropene trimer
Mixture of perfluoro-2,3,5-trimethylhexene-3,perfluoro-2,3,5-trimethylhexene-2 & perfluoro-2,4,5-trimethylhexene-2
Vzorec:
C₉F₁₈ Bod varu: 110…115 °C Hustota: 1,83 g/cm³ (25 °C) Bod vzplanutí: 24 °C Storage Temperature: Ambient |
MDL Number:
MFCD00043821 CAS číslo: 6792-31-0 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...