Tisk…

1,1,1,3,3,3-Hexafluoroisopropyltoluene-4-sulphonate 97%

Dodavatel: Apollo Scientific

Synonyms: 1,1,1,3,3,3-Hexafluoropropan-2-yl 4-methylbenzenesulphonate

PC4760-25G PC4760-5G
APOSPC4760-25GEA 4240 CZK
APOSPC4760-25G APOSPC4760-5G
1,1,1,3,3,3-Hexafluoroisopropyltoluene-4-sulphonate 97%
1,1,1,3,3,3-Hexafluoroisopropyltoluene-4-sulphonate
Vzorec: C₁₀H₈F₆O₃S
Bod tání: 39…40 °C
Storage Temperature: Ambient
MDL Number: MFCD00039262
CAS číslo: 67674-48-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR