Hexafluoroglutaryl dichloride 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC4720-5G
PC4720-25G
APOSPC4720-5GEA
3940
CZK
APOSPC4720-5G
APOSPC4720-25G
Hexafluoroglutaryl dichloride 98%
Hexafluoroglutaryl dichloride
Vzorec:
C₅Cl₂F₆O₂ MW: 276,95 g/mol Bod varu: 111 °C Hustota: 1,64 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00039309 CAS číslo: 678-77-3 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...