Hexafluoroglutaric acid 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
Hexafluoro-1,5-pentanedioic acid
PC4700-10G
PC4700-25G
PC4700-5G
APOSPC4700-10GEA
0
CZK
APOSPC4700-10G
APOSPC4700-25G
APOSPC4700-5G
Hexafluoroglutaric acid 98%
Hexafluoroglutaric acid
Vzorec:
C₅H₂F₆O₄ Bod varu: 134…138 °C/3mmHg Bod tání: 88…97 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004172 CAS číslo: 376-73-8 EINECS: 206-813-9 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...