Fluoromethyl-1,1,1,3,3,3-hexafluoroisopropyl ether
Dodavatel: Apollo Scientific
|
Varování
|
PC4681-100G
PC4681-25G
APOSPC4681-100GEA
14880
CZK
APOSPC4681-100G
APOSPC4681-25G
Fluoromethyl-1,1,1,3,3,3-hexafluoroisopropyl ether
Fluoromethyl-1,1,1,3,3,3-hexafluoroisopropyl ether
Vzorec:
C₄H₃F₇O MW: 200,06 g/mol Bod varu: 58 °C Hustota: 1,505 g/cm³ (25 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00153189 CAS číslo: 28523-86-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...