Tisk…

1H,1H,3H-Hexafluorobutyl acrylate 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: HFBA, 2,2,3,4,4,4-Hexafluorobutyl Acrylate, Acrylic acid 2,2,3,4,4,4-Hexafluorobutyl ester

PC4655-100G PC4655-5G
APOSPC4655-100GEA 2110 CZK
APOSPC4655-100G APOSPC4655-5G
1H,1H,3H-Hexafluorobutyl acrylate 95%
1H,1H,3H-Hexafluorobutyl acrylate

Stabilised; Refrigeration will extend shelf-life.

Vzorec: C₇H₆F₆O₂
MW: 236,1118 g/mol
Bod varu: 40…43 °C/8mmHg
Hustota: 1,389 g/cm³ (25 °C)
Bod vzplanutí: 60 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00044091
CAS číslo: 54052-90-3
UN: 3272
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR