5-Bromo-2-oxo-1-[3-(trifluoromethyl)benzyl]-1,2-dihydro-3-pyridine carboxylic acid 97%
Dodavatel: Apollo Scientific
PC4641-1G
PC4641-500MG
PC4641-5G
APOSPC4641-1GEA
11540
CZK
APOSPC4641-1G
APOSPC4641-500MG
APOSPC4641-5G
5-Bromo-2-oxo-1-[3-(trifluoromethyl)benzyl]-1,2-dihydro-3-pyridine carboxylic acid 97%
5-Bromo-2-oxo-1-[3-(trifluoromethyl)benzyl]-1,2-dihydro-3-pyridine carboxylic acid
Vzorec:
C₁₄H₉BrF₃NO₃ Bod tání: 187…188 °C Storage Temperature: Ambient |
MDL Number:
MFCD01935940 CAS číslo: 338982-36-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...