Tisk…

1-[(Ammoniooxy)methyl]-2-fluorobenzene chloride

Dodavatel: Apollo Scientific
Varování

Synonyms: O-(2-Fluorobenzyl)hydroxylamine hydrochloride

PC4625-5G
APOSPC4625-5GEA 3410 CZK
APOSPC4625-5G
1-[(Ammoniooxy)methyl]-2-fluorobenzene chloride
1-[(Ammoniooxy)methyl]-2-fluorobenzene chloride
Vzorec: C₇H₉ClFNO
MW: 177,6039 g/mol
Bod tání: 180…185 °C
Storage Temperature: Ambient
MDL Number: MFCD01114584
CAS číslo: 215599-91-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR