Tisk…

2'-Fluoro-6'-methoxyacetophenone

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-(2-Fluoro-6-methoxyphenyl)ethanone

PC4586-1G PC4586-25G PC4586-5G
APOSPC4586-1GEA 622 CZK
APOSPC4586-1G APOSPC4586-25G APOSPC4586-5G
2'-Fluoro-6'-methoxyacetophenone
2'-Fluoro-6'-methoxyacetophenone
Vzorec: C₉H₉FO₂
MW: 168,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00671762
CAS číslo: 120484-50-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR