2-Amino-4'-fluoroacetophenone hydrochloride 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC4576-1G
PC4576-5G
APOSPC4576-1GEA
622
CZK
APOSPC4576-1G
APOSPC4576-5G
2-Amino-4'-fluoroacetophenone hydrochloride 95%
2-Amino-4'-fluoroacetophenone hydrochloride
Vzorec:
C₈H₉ClFNO Bod tání: 238…242 °C Storage Temperature: Ambient |
MDL Number:
MFCD00193059 CAS číslo: 456-00-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...