Tisk…

2-Amino-2'-fluoroacetophenone hydrochloride 95%

Dodavatel: Apollo Scientific
Varování

PC4573-1G PC4573-5G PC4573-25G
APOSPC4573-1GEA 787 CZK
APOSPC4573-1G APOSPC4573-5G APOSPC4573-25G
2-Amino-2'-fluoroacetophenone hydrochloride 95%
2-Amino-2'-fluoroacetophenone hydrochloride
Vzorec: C₈H₉ClFNO
Bod tání: 199…203 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08532458
CAS číslo: 93102-96-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR