2-Amino-4-(trifluoromethyl)-1,3-thiazole-5-carboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Amino-4-(trifluoromethyl)thiazole-5-carboxylic Acid
PC4565-1G
PC4565-5G
PC4565-10G
APOSPC4565-1GEA
2900
CZK
APOSPC4565-1G
APOSPC4565-5G
APOSPC4565-10G
2-Amino-4-(trifluoromethyl)-1,3-thiazole-5-carboxylic acid
2-Amino-4-(trifluoromethyl)-1,3-thiazole-5-carboxylic acid
Vzorec:
C₅H₃F₃N₂O₂S MW: 212,15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00269431 CAS číslo: 239135-55-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...