3,3,3-Trifluoropropionitrile 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC4553-1G
PC4553-25G
PC4553-5G
APOSPC4553-1GEA
1870
CZK
APOSPC4553-1G
APOSPC4553-25G
APOSPC4553-5G
3,3,3-Trifluoropropionitrile 97%
3,3,3-Trifluoropropionitrile
Vzorec:
C₃H₂F₃N Bod varu: 92 °C Hustota: 1,279 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD07784277 CAS číslo: 20530-38-5 UN: 3275 ADR: 6.1+3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...