Tisk…

4-Chloro-6-fluoroquinoline

Dodavatel: Apollo Scientific
Varování

PC4517-1G PC4517-250MG
APOSPC4517-1GEA 705 CZK
APOSPC4517-1G APOSPC4517-250MG
4-Chloro-6-fluoroquinoline
4-Chloro-6-fluoroquinoline
Vzorec: C₉H₅ClFN
MW: 181,60 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00278783
CAS číslo: 391-77-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR