Tisk…

Bis(2,2,2-trifluoroethyl) Carbonate

Dodavatel: Apollo Scientific
Varování

Synonyms: Carbonic acid bis(2,2,2-trifluoroethyl) ester

PC450295-25G
APOSPC450295-25GEA 3910 CZK
APOSPC450295-25G
Bis(2,2,2-trifluoroethyl) Carbonate
Bis(2,2,2-trifluoroethyl) Carbonate
Vzorec: C₅H₄F₆O₃
MW: 226,07 g/mol
Bod varu: 118 °C
Hustota: 1,51 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD11857845
CAS číslo: 1513-87-7
UN: 3272
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR