Tisk…

2-Chlorophenyl trifluoromethanesulphonate

Dodavatel: Apollo Scientific

Synonyms: 2-Chlorophenyl Triflate, Trifluoromethanesulphonic Acid 2-Chlorophenyl Ester

PC450239-1G PC450239-25G PC450239-5G
APOSPC450239-1GEA 2280 CZK
APOSPC450239-1G APOSPC450239-25G APOSPC450239-5G
2-Chlorophenyl trifluoromethanesulphonate
2-Chlorophenyl trifluoromethanesulphonate
Vzorec: C₇H₄ClF₃O₃S
MW: 260,61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07784322
CAS číslo: 66107-36-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR