Ethyl-2-amino-4,4,4-trifluoro-3-(trifluoromethyl)butyrate
Dodavatel: Apollo Scientific
Synonyms:
Ethyl 4,4,4,4',4',4'-hexafluoro-DL-valinate, 4,4,4,4',4',4'-Hexafluorovaline, ethyl ester
PC450233-1G
PC450233-5G
APOSPC450233-1GEA
2240
CZK
APOSPC450233-1G
APOSPC450233-5G
Ethyl-2-amino-4,4,4-trifluoro-3-(trifluoromethyl)butyrate
Ethyl-2-amino-4,4,4-trifluoro-3-(trifluoromethyl)butyrate
Vzorec:
C₇H₉F₆NO₂ MW: 253,15 g/mol Bod varu: 70…72 °C/10mmHg Storage Temperature: Ambient |
MDL Number:
MFCD00040845 CAS číslo: 78185-92-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...