Tisk…

2-Naphthyl trifluoromethanesulphonate

Dodavatel: Apollo Scientific

PC450164-25G PC450164-5G
APOSPC450164-25GEA 11410 CZK
APOSPC450164-25G APOSPC450164-5G
2-Naphthyl trifluoromethanesulphonate
2-Naphthyl trifluoromethanesulphonate
Vzorec: C₁₁H₇F₃O₃S
Storage Temperature: Ambient
MDL Number: MFCD00192341
CAS číslo: 3857-83-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR