Tisk…
1-Methyl-2-trifluoro-methyl benzimidazole
Dodavatel: Apollo Scientific
Synonyms: 1H-Benzimidazole, 1-Methyl-2-(trifluoromethyl)-1H-benzo[d]imidazole
PC450146-1G PC450146-5G
APOSPC450146-1GEA 5000 CZK
APOSPC450146-1G APOSPC450146-5G
1-Methyl-2-trifluoro-methyl benzimidazole
1-Methyl-2-trifluoro-methyl benzimidazole
Vzorec: C₉H₇F₃N₂
MW: 200.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00561648
CAS číslo: 384-46-3