Tisk…

N-[4-Bromo-2-nitro-5-(trifluoromethyl)phenyl]-2,2,2-trifluoroacetamide

Dodavatel: Apollo Scientific

Synonyms: 2-Bromo-4-nitro-5-(trifluoroacetamido)benzotrifluoride

PC450105-1G PC450105-5G
APOSPC450105-1GEA 3940 CZK
APOSPC450105-1G APOSPC450105-5G
N-[4-Bromo-2-nitro-5-(trifluoromethyl)phenyl]-2,2,2-trifluoroacetamide
N-[4-Bromo-2-nitro-5-(trifluoromethyl)phenyl]-2,2,2-trifluoroacetamide
Vzorec: C₉H₃BrF₆N₂O₃
MW: 381,04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14584783
CAS číslo: 156425-51-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR