3-Bromo-2,6-difluorophenol
Dodavatel: Apollo Scientific
|
Varování
|
PC450015-1G
PC450015-5G
APOSPC450015-1GEA
2440
CZK
APOSPC450015-1G
APOSPC450015-5G
3-Bromo-2,6-difluorophenol
3-Bromo-2,6-difluorophenol
Vzorec:
C₆H₃BrF₂O MW: 208.9882 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD12910487 CAS číslo: 221220-99-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...