Tisk…

2,4-Dichloro-6-fluoroquinoline

Dodavatel: Apollo Scientific
Varování

Synonyms: 2,4-Dichloro-6-fluoro-quinoline

PC449020-1G PC449020-5G PC449020-25G
APOSPC449020-1GEA 2440 CZK
APOSPC449020-1G APOSPC449020-5G APOSPC449020-25G
2,4-Dichloro-6-fluoroquinoline
2,4-Dichloro-6-fluoroquinoline
Vzorec: C₉H₄Cl₂FN
MW: 216.0392 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09261383
CAS číslo: 406204-74-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR