1-Cyclopropyl-4,4,4-trifluoro-1,3-butanedione
Dodavatel: Apollo Scientific
PC449003-1G
PC449003-5G
PC449003-250MG
APOSPC449003-1GEA
0
CZK
APOSPC449003-1G
APOSPC449003-5G
APOSPC449003-250MG
1-Cyclopropyl-4,4,4-trifluoro-1,3-butanedione
1-Cyclopropyl-4,4,4-trifluoro-1,3-butanedione
Vzorec:
C₇H₇F₃O₂ Storage Temperature: Ambient |
MDL Number:
MFCD03419771 CAS číslo: 30923-69-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...