Tisk…

4-(Trifluoromethoxy)phenyl isocyanate 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-Isocyanato-4-(trifluoromethoxy)benzene

PC4489-25G PC4489-5G
APOSPC4489-25GEA 2070 CZK
APOSPC4489-25G APOSPC4489-5G
4-(Trifluoromethoxy)phenyl isocyanate 95%
4-(Trifluoromethoxy)phenyl isocyanate

  • May form small amount of sediment.

Vzorec: C₈H₄F₃NO₂
MW: 203,12 g/mol
Bod varu: 78 °C/22mmHg
Hustota: 1,346 g/cm³ (25 °C)
Bod vzplanutí: 70 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00035702
CAS číslo: 35037-73-1
EINECS: 252-328-0
UN: 2206
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR