Tisk…

2-Bromo-2',6'-dichloro-3'-fluoroacetophenone 96%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2,6-Dichloro-3-fluorophenacyl bromide

PC4487-1G PC4487-5G
APOSPC4487-1GEA 7610 CZK
APOSPC4487-1G APOSPC4487-5G
2-Bromo-2',6'-dichloro-3'-fluoroacetophenone 96%
2-Bromo-2',6'-dichloro-3'-fluoroacetophenone
Vzorec: C₈H₄BrCl₂FO
Bod tání: 42…44 °C
Storage Temperature: Ambient
MDL Number: MFCD09038425
CAS číslo: 914635-49-7
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR