Tisk…

1-Chloro-2,3-difluorobenzene

Dodavatel: Apollo Scientific
Varování

PC4486-1G PC4486-25G PC4486-5G
APOSPC4486-1GEA 622 CZK
APOSPC4486-1G APOSPC4486-25G APOSPC4486-5G
1-Chloro-2,3-difluorobenzene
1-Chloro-2,3-difluorobenzene
Vzorec: C₆H₃ClF₂
MW: 148,5400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04112584
CAS číslo: 36556-47-5
UN: 1993
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR