Heptafluorobutyric anhydride (HFAA) 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
HFBA, 2,2,3,3,4,4,4-Heptafluorobutanoic anhydride, Perfluorobutyric anhydride, HFAA
PC4470-100G
PC4470-25G
APOSPC4470-100GEA
8500
CZK
APOSPC4470-100G
APOSPC4470-25G
Heptafluorobutyric anhydride (HFAA) 95%
Heptafluorobutyric anhydride
Useful for derivitisation of amines, alcohols, amino acids and other bioactive compounds for GC analysis.
Vzorec:
C₈F₁₄O₃ MW: 410,06 g/mol Bod varu: 106…107 °C Bod tání: –43 °C Hustota: 1,665 g/cm³ (25 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000432 CAS číslo: 336-59-4 EINECS: 206-410-8 UN: 3265 ADR: 8,II |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...