Tisk…

4-Bromo-2,6-difluorobenzamide 95+%

Dodavatel: Apollo Scientific

PC446191-1G
APOSPC446191-1GEA 8600 CZK
APOSPC446191-1G
4-Bromo-2,6-difluorobenzamide 95+%
4-Bromo-2,6-difluorobenzamide
Vzorec: C₇H₄BrF₂NO
MW: 236,014 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13689151
CAS číslo: 840481-49-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR