Tisk…

4-Bromo-3-fluorobenzamide 95+%

Dodavatel: Apollo Scientific

PC446147-1G PC446147-5G
APOSPC446147-1GEA 2690 CZK
APOSPC446147-1G APOSPC446147-5G
4-Bromo-3-fluorobenzamide 95+%
4-Bromo-3-fluorobenzamide
Vzorec: C₇H₅BrFNO
MW: 218,025 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06409121
CAS číslo: 759427-20-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR