[3-Chloro-5-trifluoromethyl-1H-pyridin-(2E)-ylidene]cyanoacetic acid ethyl ester
Dodavatel: Apollo Scientific
PC446064-1G
PC446064-5G
APOSPC446064-1GEA
6950
CZK
APOSPC446064-1G
APOSPC446064-5G
[3-Chloro-5-trifluoromethyl-1H-pyridin-(2E)-ylidene]cyanoacetic acid ethyl ester
[3-Chloro-5-trifluoromethyl-1H-pyridin-(2E)-ylidene]cyanoacetic acid ethyl ester
Vzorec:
C₁₁H₈ClF₃N₂O₂ MW: 292.65 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18384848 CAS číslo: 1300019-72-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...