[5-(Trifluoromethyl)-1-benzothiophen-2-yl]methanol
Dodavatel: Apollo Scientific
|
Varování
|
PC446045-1G
PC446045-250MG
APOSPC446045-1GEA
2820
CZK
APOSPC446045-1G
APOSPC446045-250MG
[5-(Trifluoromethyl)-1-benzothiophen-2-yl]methanol
[5-(Trifluoromethyl)-1-benzothiophen-2-yl]methanol
Vzorec:
C₁₀H₇F₃OS MW: 232,23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11841028 CAS číslo: 951122-90-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...